siRNA Details
| Gene Symbol: Bpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004079.1 | NP_001004079.1 | 8, 9 | 1129 | ||
| XM_008762323.1 | XP_008760545.1 | 8, 9 | 1139 | |||
| GenBank mRNA | BC079318.1 | - | 8, 9 | 1129 | 7135061 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpi| Gene Symbol: Bpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004079.1 | NP_001004079.1 | 8, 9 | 1129 | ||
| XM_008762323.1 | XP_008760545.1 | 8, 9 | 1139 | |||
| GenBank mRNA | BC079318.1 | - | 8, 9 | 1129 | 7135061 | |