siRNA Details
| Gene Symbol: ZNF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017396.2 | NP_001017396.1 | 3 | 631 | ||
| NM_001291605.1 | NP_001278534.1 | 4 | 703 | |||
| NM_021088.3 | NP_066574.2 | 4 | 703 | |||
| GenBank mRNA | AK055072.1 | - | 4 | 682 | ||
| AK290798.1 | - | 3 | 241 | |||
| AK299058.1 | - | 4 | 303 | |||
| AK315762.1 | - | 4 | 315 | |||
| AK315829.1 | - | 3 | 236 | |||
| BC005068.1 | - | 4 | 675 | 3350363 | ||
| BC009976.2 | - | 2 | 89 | |||
| X60152.1 | - | 3 | 616 | |||
| X60154.1 | - | 4 | 292 | |||

ZNF2
Genomic Map