siRNA Details
| Gene Symbol: ZNF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017396.2 | NP_001017396.1 | 3, 4 | 688 | ||
| NM_021088.3 | NP_066574.2 | 4, 5 | 760 | |||
| GenBank mRNA | AK055072.1 | - | 4, 5 | 739 | ||
| AK290798.1 | - | 3, 4 | 298 | |||
| AK315762.1 | - | 4, 5 | 372 | |||
| AK315829.1 | - | 3, 4 | 293 | |||
| BC005068.1 | - | 4, 5 | 732 | 3350363 | ||
| BC009976.2 | - | 2, 3 | 146 | |||
| X60152.1 | - | 3, 4 | 673 | |||
| X60154.1 | - | 4, 5 | 349 | |||

ZNF2
Genomic Map