siRNA Details
| Gene Symbol: LOC103690143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762549.1 | XP_008760771.1 | 2 | 999 | ||
| Gene Symbol: Svs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102417.1 | NP_001095887.1 | 696 | |||

LOC103690143
Genomic Map