siRNA Details
| Gene Symbol: Pigt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106540.2 | NP_001100010.2 | 9, 10 | 1267 | ||
| XM_006235532.2 | XP_006235594.1 | 9, 10 | 2556 | |||
| GenBank mRNA | FQ220416.1 | - | 2, 3 | 258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigt| Gene Symbol: Pigt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106540.2 | NP_001100010.2 | 9, 10 | 1267 | ||
| XM_006235532.2 | XP_006235594.1 | 9, 10 | 2556 | |||
| GenBank mRNA | FQ220416.1 | - | 2, 3 | 258 | ||