siRNA Details
| Gene Symbol: Ctsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011959.1 | NP_001011959.1 | 1, 2 | 340 | ||
| XM_006235534.1 | XP_006235596.1 | 2, 3 | 598 | |||
| GenBank mRNA | BC078934.1 | - | 1, 2 | 340 | 7108104 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsa| Gene Symbol: Ctsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011959.1 | NP_001011959.1 | 1, 2 | 340 | ||
| XM_006235534.1 | XP_006235596.1 | 2, 3 | 598 | |||
| GenBank mRNA | BC078934.1 | - | 1, 2 | 340 | 7108104 | |