siRNA Details
| Gene Symbol: Pltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168543.1 | NP_001162015.1 | 8 | 943 | ||
| XM_006235535.2 | XP_006235597.1 | 9 | 1001 | |||
| XM_008762476.1 | XP_008760698.1 | 9 | 995 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pltp| Gene Symbol: Pltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168543.1 | NP_001162015.1 | 8 | 943 | ||
| XM_006235535.2 | XP_006235597.1 | 9 | 1001 | |||
| XM_008762476.1 | XP_008760698.1 | 9 | 995 | |||