siRNA Details
| Gene Symbol: Ncoa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106543.1 | NP_001100013.1 | 3 | 321 | ||
| XM_008762477.1 | XP_008760699.1 | 2 | 474 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncoa5| Gene Symbol: Ncoa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106543.1 | NP_001100013.1 | 3 | 321 | ||
| XM_008762477.1 | XP_008760699.1 | 2 | 474 | |||