siRNA Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191744.1 | NP_001178673.1 | 5 | 761 | ||
| XM_006235679.2 | XP_006235741.1 | 5 | 1670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cass4| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191744.1 | NP_001178673.1 | 5 | 761 | ||
| XM_006235679.2 | XP_006235741.1 | 5 | 1670 | |||