siRNA Details
| Gene Symbol: Rtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033890.1 | NP_001029062.1 | 8 | 770 | ||
| GenBank mRNA | BC101880.1 | - | 8 | 770 | 7929941 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtfdc1| Gene Symbol: Rtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033890.1 | NP_001029062.1 | 8 | 770 | ||
| GenBank mRNA | BC101880.1 | - | 8 | 770 | 7929941 | |