siRNA Details
| Gene Symbol: Itgav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762000.1 | XP_008760222.1 | 24 | 3316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgav| Gene Symbol: Itgav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762000.1 | XP_008760222.1 | 24 | 3316 | ||