siRNA Details
| Gene Symbol: Itgav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762000.1 | XP_008760222.1 | 24 | 3242 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008402, AK036485, U14135, AK154307, AK149984.

Itgav
Genomic Map