siRNA Details
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013079.1 | NP_001013097.1 | 7 | 659 | ||
| XM_006235816.2 | XP_006235878.1 | 8 | 908 | |||
| GenBank mRNA | BC091208.1 | - | 7 | 659 | 7381353 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl2| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013079.1 | NP_001013097.1 | 7 | 659 | ||
| XM_006235816.2 | XP_006235878.1 | 8 | 908 | |||
| GenBank mRNA | BC091208.1 | - | 7 | 659 | 7381353 | |