siRNA Details
| Gene Symbol: Ythdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024756.1 | NP_001019927.1 | 4 | 1598 | ||
| GenBank mRNA | BC097360.1 | - | 4 | 1598 | 7442371 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ythdf1| Gene Symbol: Ythdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024756.1 | NP_001019927.1 | 4 | 1598 | ||
| GenBank mRNA | BC097360.1 | - | 4 | 1598 | 7442371 | |