siRNA Details
| Gene Symbol: Nkain4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106550.1 | NP_001100020.1 | 2 | 279 | ||
| XM_006235744.2 | XP_006235806.1 | 4 | 533 | |||
| XM_006235745.2 | XP_006235807.1 | 4 | 2415 | |||
| XM_008762496.1 | XP_008760718.1 | 5 | 882 | |||
| XM_008762497.1 | XP_008760719.1 | 5 | 1710 | |||
| XM_008762498.1 | XP_008760720.1 | 5 | 685 | |||

Nkain4
Genomic Map