siRNA Details
| Gene Symbol: ZNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021089.2 | NP_066575.2 | 4 | 925 | ||
| GenBank mRNA | AK093110.1 | - | 4 | 925 | ||
| AK225825.1 | - | 4 | 921 | |||
| AK299716.1 | - | 2 | 291 | |||
| BC039323.1 | - | 4 | 883 | 5267031 | ||
| M29581.1 | - | 3 | 699 | |||

ZNF8
Genomic Map