siRNA Details
| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011961.1 | NP_001011961.1 | 1, 2 | 496 | ||
| GenBank mRNA | BC090006.1 | - | 1, 2 | 496 | 7323399 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srms| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011961.1 | NP_001011961.1 | 1, 2 | 496 | ||
| GenBank mRNA | BC090006.1 | - | 1, 2 | 496 | 7323399 | |