siRNA Details
| Gene Symbol: Dlx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191746.1 | NP_001178675.1 | 1, 2 | 407 | ||
| GenBank mRNA | S81930.1 | - | 1, 2 | 224 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx2| Gene Symbol: Dlx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191746.1 | NP_001178675.1 | 1, 2 | 407 | ||
| GenBank mRNA | S81930.1 | - | 1, 2 | 224 | ||