siRNA Details
| Gene Symbol: Sp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106552.1 | NP_001100022.1 | 2 | 816 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp5| Gene Symbol: Sp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106552.1 | NP_001100022.1 | 2 | 816 | ||