siRNA Details
| Gene Symbol: ZNF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015394.4 | NP_056209.2 | 5 | 1276 | ||
| GenBank mRNA | AK314784.1 | - | 5 | 1537 | ||
| AL110188.1 | - | 3 | 993 | |||
| BC024182.2 | - | 5 | 1277 | 4823159 | ||
| X52332.1 | - | 5 | 758 | |||

ZNF10
Genomic Map