siRNA Details
| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013080.2 | NP_001013098.2 | 5 | 502 | ||
| XM_006233590.2 | XP_006233652.1 | 5 | 484 | |||
| XM_008761584.1 | XP_008759806.1 | 4 | 1190 | |||
| XM_008761585.1 | XP_008759807.1 | 5 | 1145 | |||
| GenBank mRNA | BC089933.1 | - | 2 | 828 | ||
| CF115110.1 | - | 5 | 502 | |||

Clic3
Genomic Map