siRNA Details
| Gene Symbol: Camsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168549.1 | NP_001162021.1 | 12 | 3859 | ||
| XM_006233672.2 | XP_006233734.1 | 12 | 3827 | |||
| XM_006233673.2 | XP_006233735.1 | 11 | 3794 | |||
| XM_006233674.2 | XP_006233736.1 | 11 | 3794 | |||
| XM_006233675.2 | XP_006233737.1 | 12 | 3827 | |||
| XM_006233677.2 | XP_006233739.1 | 7 | 2739 | |||
| GenBank mRNA | FQ211685.1 | - | 1 | 518 | ||

Camsap1
Genomic Map