siRNA Details
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106561.1 | NP_001100031.1 | 6 | 1355 | ||
| XM_006233756.2 | XP_006233818.1 | 4 | 397 | |||
| XM_008761633.1 | XP_008759855.1 | 3 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacfd1| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106561.1 | NP_001100031.1 | 6 | 1355 | ||
| XM_006233756.2 | XP_006233818.1 | 4 | 397 | |||
| XM_008761633.1 | XP_008759855.1 | 3 | 271 | |||