siRNA Details
| Gene Symbol: Slc2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106562.1 | NP_001100032.1 | 6 | 913 | ||
| XM_006233757.2 | XP_006233819.1 | 6 | 904 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a6| Gene Symbol: Slc2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106562.1 | NP_001100032.1 | 6 | 913 | ||
| XM_006233757.2 | XP_006233819.1 | 6 | 904 | |||