siRNA Details
| Gene Symbol: Vav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106563.2 | NP_001100033.2 | 23 | 2531 | ||
| XM_006233761.2 | XP_006233823.1 | 23 | 2597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vav2| Gene Symbol: Vav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106563.2 | NP_001100033.2 | 23 | 2531 | ||
| XM_006233761.2 | XP_006233823.1 | 23 | 2597 | |||