siRNA Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106565.1 | NP_001100035.1 | 6 | 758 | ||
| XM_008761638.1 | XP_008759860.1 | 6 | 764 | |||
| XM_008761639.1 | XP_008759861.1 | 5 | 797 | |||
| GenBank mRNA | BC167032.1 | - | 6 | 733 | 9092805 | |

Med27
Genomic Map