siRNA Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106565.1 | NP_001100035.1 | 2 | 373 | ||
| XM_006233767.2 | XP_006233829.1 | 2 | 496 | |||
| XM_008761638.1 | XP_008759860.1 | 2 | 379 | |||
| GenBank mRNA | BC167032.1 | - | 2 | 348 | 9092805 | |

Med27
Genomic Map