siRNA Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013081.1 | NP_001013099.1 | 6, 7 | 468 | ||
| XM_006233777.2 | XP_006233839.1 | 7, 8 | 852 | |||
| XM_006233778.2 | XP_006233840.1 | 7, 8 | 852 | |||
| XM_006233779.2 | XP_006233841.1 | 7, 8 | 1545 | |||
| XM_006233780.2 | XP_006233842.1 | 7, 8 | 608 | |||
| GenBank mRNA | BC091389.1 | - | 6, 7 | 468 | 7321523 | |
| FQ212963.1 | - | 6, 7 | 484 | |||
| FQ219402.1 | - | 6, 7 | 489 | |||
| FQ219572.1 | - | 3, 4 | 471 | |||
| FQ223356.1 | - | 6, 7 | 475 | |||

Phyhd1
Genomic Map