siRNA Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106567.1 | NP_001100037.1 | 5 | 643 | ||
| XM_006233783.2 | XP_006233845.1 | 5 | 1680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dolpp1| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106567.1 | NP_001100037.1 | 5 | 643 | ||
| XM_006233783.2 | XP_006233845.1 | 5 | 1680 | |||