siRNA Details
| Gene Symbol: Ppapdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012349.2 | NP_001012349.1 | 2 | 928 | ||
| GenBank mRNA | BC089948.1 | - | 2 | 928 | 7311891 | |
| CB771354.1 | - | 1 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppapdc3| Gene Symbol: Ppapdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012349.2 | NP_001012349.1 | 2 | 928 | ||
| GenBank mRNA | BC089948.1 | - | 2 | 928 | 7311891 | |
| CB771354.1 | - | 1 | 276 | |||