siRNA Details
| Gene Symbol: Ciz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106568.1 | NP_001100038.1 | 13 | 2411 | ||
| XM_006233940.2 | XP_006234002.1 | 13 | 2019 | |||
| XM_006233941.2 | XP_006234003.1 | 13 | 1979 | |||
| XM_006233943.2 | XP_006234005.1 | 13 | 1964 | |||
| XM_006233944.1 | XP_006234006.1 | 12 | 1952 | |||
| XM_006233946.2 | XP_006234008.1 | 14 | 1866 | |||
| XM_008761714.1 | XP_008759936.1 | 12 | 2396 | |||
| XM_008761715.1 | XP_008759937.1 | 12 | 2381 | |||
| XM_008761716.1 | XP_008759938.1 | 11 | 2324 | |||
| XM_008761717.1 | XP_008759939.1 | 13 | 2254 | |||
| XM_008761718.1 | XP_008759940.1 | 13 | 2242 | |||
| XM_008761719.1 | XP_008759941.1 | 13 | 2227 | |||
| GenBank mRNA | BC166776.1 | - | 14 | 1788 | 7934431 | |

Ciz1
Genomic Map