siRNA Details
| Gene Symbol: Olr397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000380.1 | NP_001000380.1 | 1 | 94 | ||
| XM_006234064.2 | XP_006234126.1 | 2 | 431 | |||
| XM_006234065.2 | XP_006234127.1 | 2 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr397| Gene Symbol: Olr397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000380.1 | NP_001000380.1 | 1 | 94 | ||
| XM_006234064.2 | XP_006234126.1 | 2 | 431 | |||
| XM_006234065.2 | XP_006234127.1 | 2 | 428 | |||