siRNA Details
| Gene Symbol: ZNF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006959.2 | NP_008890.2 | 3 | 1600 | ||
| XM_017027205.1 | XP_016882694.1 | 4 | 3550 | |||
| GenBank mRNA | AB075827.1 | - | 1 | 1207 | ||
| AK097123.1 | - | 1 | 886 | |||
| AK098183.1 | - | 3 | 1582 | |||
| AK127948.1 | - | 4 | 1639 | |||
| AK128471.1 | - | 1 | 1805 | |||
| X52341.1 | - | 1 | 46 | |||

ZNF17
Genomic Map