siRNA Details
| Gene Symbol: Lhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106571.1 | NP_001100041.1 | 5 | 1032 | ||
| XM_006234072.2 | XP_006234134.1 | 5 | 1166 | |||
| XM_006234073.1 | XP_006234135.1 | 5 | 905 | |||
| XM_008761742.1 | XP_008759964.1 | 5 | 1181 | |||

Lhx2
Genomic Map