siRNA Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013925.1 | NP_001013947.1 | 3 | 337 | ||
| GenBank mRNA | BC083586.1 | - | 3 | 337 | 7131376 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nub1| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013925.1 | NP_001013947.1 | 3 | 337 | ||
| GenBank mRNA | BC083586.1 | - | 3 | 337 | 7131376 | |