siRNA Details
| Gene Symbol: Smarcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011966.1 | NP_001011966.1 | 12 | 1324 | ||
| XM_006235883.2 | XP_006235945.1 | 11 | 1421 | |||
| XM_006235884.2 | XP_006235946.1 | 11 | 1421 | |||
| XM_006235885.2 | XP_006235947.1 | 11 | 1385 | |||
| XM_006235886.2 | XP_006235948.1 | 12 | 1408 | |||
| XM_006235887.2 | XP_006235949.1 | 12 | 1372 | |||
| XM_006235888.2 | XP_006235950.1 | 11 | 1421 | |||
| XM_006235889.2 | XP_006235951.1 | 11 | 1385 | |||
| GenBank mRNA | BC085349.1 | - | 12 | 1324 | 7107899 | |

Smarcd3
Genomic Map