siRNA Details
| Gene Symbol: Srpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106575.2 | NP_001100045.1 | 10 | 1460 | ||
| XM_006235903.2 | XP_006235965.1 | 10 | 4456 | |||
| XM_008762635.1 | XP_008760857.1 | 10 | 3878 | |||
| XM_008762636.1 | XP_008760858.1 | 10 | 4429 | |||
| XM_008762637.1 | XP_008760859.1 | 10 | 4455 | |||
| GenBank mRNA | BC161879.1 | - | 10 | 1460 | 7125533 | |

Srpk2
Genomic Map