siRNA Details
| Gene Symbol: Sema3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106578.1 | NP_001100048.1 | 5 | 490 | ||
| XM_008762645.1 | XP_008760867.1 | 6 | 799 | |||
| XM_008762646.1 | XP_008760868.1 | 6 | 797 | |||
| GenBank mRNA | BC169108.1 | - | 5 | 490 | 7096896 | |

Sema3c
Genomic Map