siRNA Details
Gene Symbol: LOC102551166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235984.2 | XP_006236046.1 | 6 | 904 | ||
XM_006235985.2 | XP_006236047.1 | 6 | 904 | |||
XM_006235986.2 | XP_006236048.1 | 6 | 904 | |||
XM_006235987.2 | XP_006236049.1 | 2 | 134 |
