siRNA Details
Gene Symbol: Lrrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_216085.7 | XP_216085.3 | 2 | 1467 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_216085.7 | XP_216085.3 | 2 | 1467 |