siRNA Details
| Gene Symbol: Las1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170590.1 | NP_001164061.1 | 10 | 1522 | ||
| XM_008773298.1 | XP_008771520.1 | 10 | 1513 | |||
| GenBank mRNA | BC169024.1 | - | 3 | 1035 | 9029735 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Las1l| Gene Symbol: Las1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170590.1 | NP_001164061.1 | 10 | 1522 | ||
| XM_008773298.1 | XP_008771520.1 | 10 | 1513 | |||
| GenBank mRNA | BC169024.1 | - | 3 | 1035 | 9029735 | |