siRNA Details
| Gene Symbol: ST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004102.1 | NP_001004102.1 | 3 | 429 | ||
| XM_006236132.2 | XP_006236194.1 | 3 | 1269 | |||
| XM_006236133.2 | XP_006236195.1 | 3 | 1562 | |||
| XM_006236134.2 | XP_006236196.1 | 3 | 798 | |||
| XM_006236135.2 | XP_006236197.1 | 3 | 798 | |||
| XM_008762700.1 | XP_008760922.1 | 3 | 1994 | |||
| XM_008762701.1 | XP_008760923.1 | 4 | 956 | |||
| GenBank mRNA | BC078945.1 | - | 5 | 685 | ||
| BC099820.1 | - | 3 | 511 | 7930680 | ||

ST7
Genomic Map