siRNA Details
| Gene Symbol: Lmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100964.1 | NP_001094434.1 | 2 | 969 | ||
| GenBank mRNA | AB331240.1 | - | 2 | 845 | ||
| BC128755.1 | - | 2 | 969 | 7104774 | ||
| Gene Symbol: LOC100909784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749707.3 | XP_003749755.1 | 2 | 1041 | ||

Lmod2
Genomic Map