siRNA Details
| Gene Symbol: Iqub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034130.1 | NP_001029302.1 | 11 | 1823 | ||
| GenBank mRNA | BC099086.1 | - | 11 | 1823 | 7454794 | |
| DQ132434.1 | - | 11 | 1870 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqub| Gene Symbol: Iqub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034130.1 | NP_001029302.1 | 11 | 1823 | ||
| GenBank mRNA | BC099086.1 | - | 11 | 1823 | 7454794 | |
| DQ132434.1 | - | 11 | 1870 | |||