siRNA Details
| Gene Symbol: Kcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762841.1 | XP_008761063.1 | 36, 37 | 5464 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcp| Gene Symbol: Kcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762841.1 | XP_008761063.1 | 36, 37 | 5464 | ||