siRNA Details
| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106587.1 | NP_001100057.1 | 4 | 851 | ||
| XM_006236186.2 | XP_006236248.1 | 5 | 625 | |||
| GenBank mRNA | AY724521.1 | - | 4 | 625 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnpo3| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106587.1 | NP_001100057.1 | 4 | 851 | ||
| XM_006236186.2 | XP_006236248.1 | 5 | 625 | |||
| GenBank mRNA | AY724521.1 | - | 4 | 625 | ||