siRNA Details
| Gene Symbol: Zc3hc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127293.1 | NP_001120765.1 | 1 | 136 | ||
| GenBank mRNA | BC166834.1 | - | 1 | 136 | 9022075 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3hc1| Gene Symbol: Zc3hc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127293.1 | NP_001120765.1 | 1 | 136 | ||
| GenBank mRNA | BC166834.1 | - | 1 | 136 | 9022075 | |