siRNA Details
| Gene Symbol: ZNF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006963.4 | NP_008894.2 | 2 | 390 | ||
| GenBank mRNA | AK291508.1 | - | 2 | 292 | ||
| AK312278.1 | - | 2 | 292 | |||
| BC010642.1 | - | 2 | 280 | 3852749 | ||
| BC041139.1 | - | 2 | 741 | 5751460 | ||
| BC053687.1 | - | 2 | 284 | 5214077 | ||
| BT009806.1 | - | 1 | 147 | |||
| S50223.1 | - | 1 | 236 | |||

ZNF22
Genomic Map