siRNA Details
| Gene Symbol: Bpgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199382.1 | NP_955414.1 | 2 | 186 | ||
| XM_006236259.2 | XP_006236321.1 | 2 | 300 | |||
| GenBank mRNA | AY325190.1 | - | 2 | 95 | ||
| BC062240.1 | - | 2 | 186 | 6887677 | ||

Bpgm
Genomic Map