siRNA Details
| Gene Symbol: Atp6v0a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106591.1 | NP_001100061.1 | 15 | 1887 | ||
| XM_006236296.2 | XP_006236358.1 | 16 | 5372 | |||
| XM_006236299.2 | XP_006236361.1 | 16 | 1960 | |||
| XM_008762769.1 | XP_008760991.1 | 16 | 1925 | |||

Atp6v0a4
Genomic Map